missing translation for 'onlineSavingsMsg'
Learn More

RAB35 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16898323 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16898323 150 μL 150µL
16808333 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16898323 Supplier Proteintech Supplier No. 113292AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAB35 possesses GTPase activity.

Specifications

Antigen RAB35
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAB35
Gene Accession No. Q15286, Q5U316, Q6PHN9
Gene Alias GTP binding protein RAY, H ray, RAB1C, RAB35, Ras related protein Rab 1C, Ras related protein Rab 35, RAY
Gene Symbols Rab35
Host Species Rabbit
Immunogen RAB35 Fusion Protein Ag1872
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11021, 288700, 77407
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.