missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB35 Mouse, anti-Human, Clone: 3H3E1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RAB35 |
|---|---|
| Clone | 3H3E1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18034755
|
Proteintech
68552-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18065275
|
Proteintech
68552-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RAB35 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB35. This antibody recognizes human, mouse, rat, rabbit antigen. The RAB35 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RAB35 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat, Rabbit | |
| GTP binding protein RAY, H ray, RAB1C, RAB35, Ras related protein Rab 1C, Ras related protein Rab 35, RAY | |
| Primary |
| 3H3E1 | |
| Western Blot, ELISA | |
| Mouse | |
| 11021 | |
| IgG2a | |
| Store at -20°C. Stable for one year after shipment. |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel