Learn More
RAB35 Mouse, anti-Human, Clone: 3H3E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68552-1-IG-20UL
Description
The RAB35 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RAB35. This antibody recognizes human, mouse, rat, rabbit antigen. The RAB35 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RAB35 | |
| 3H3E1 | |
| GTP binding protein RAY, H ray, RAB1C, RAB35, Ras related protein Rab 1C, Ras related protein Rab 35, RAY | |
| 20 μL | |
| 11021 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Rabbit | |
| 150 kDa | |
| IgG2a |