missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB34 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £366.00
Specifications
| Antigen | RAB34 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18628431
|
Novus Biologicals
NBP2-93447-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668521
|
Novus Biologicals
NBP2-93447-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAB34 Polyclonal antibody specifically detects RAB34 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RAB34 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 83871 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| RAB34, member RAS oncogene family, RAHRAB39Ras-related protein Rab-39, ras-related protein Rab-34, Ras-related protein Rah | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-259 of human RAB34 (NP_114140.4). MNILAPVRRDRVLAELPQCLRKEAALHGHKDFHPRVTCACQEHRTGTVGFKISKVIVVGDLSVGKTCLINRFCKDTFDKNYKATIGVDFEMERFEVLGIPFSLQLWDTAGQERFKCIASTYYRGAQAIIIVFNLNDVASLEHTKQWLADALKENDPSSVLLFLVGSKKDLSTPAQYALMEKDALQVAQEMKAEYWAVSSLTGENVREFFFRVAALTFEANVLAELEKSGARRIGDVVRINSDDSNLYLTASKKKPTCCP | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title