missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB34 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93447-0.02ml
This item is not returnable.
View return policy
Description
RAB34 Polyclonal antibody specifically detects RAB34 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RAB34 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| RAB34, member RAS oncogene family, RAHRAB39Ras-related protein Rab-39, ras-related protein Rab-34, Ras-related protein Rah | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-259 of human RAB34 (NP_114140.4). MNILAPVRRDRVLAELPQCLRKEAALHGHKDFHPRVTCACQEHRTGTVGFKISKVIVVGDLSVGKTCLINRFCKDTFDKNYKATIGVDFEMERFEVLGIPFSLQLWDTAGQERFKCIASTYYRGAQAIIIVFNLNDVASLEHTKQWLADALKENDPSSVLLFLVGSKKDLSTPAQYALMEKDALQVAQEMKAEYWAVSSLTGENVREFFFRVAALTFEANVLAELEKSGARRIGDVVRINSDDSNLYLTASKKKPTCCP | |
| 0.02 mL | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 83871 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction