missing translation for 'onlineSavingsMsg'
Learn More

Rab27a Antibody (2E8), Novus Biologicals™

Rabbit Monoclonal Antibody

£260.00 - £406.00

Specifications

Antigen Rab27a
Clone 2E8
Dilution Flow Cytometry 1:50-1:200, ELISA
Applications Flow Cytometry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30035499
View Documents
Novus Biologicals
NBP3-26353-50ul
50 μL
£260.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30032022
View Documents
Novus Biologicals
NBP3-26353-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rab27a Monoclonal antibody specifically detects Rab27a in Human samples. It is validated for Flow Cytometry, ELISA
Specifications

Specifications

Rab27a
Flow Cytometry 1:50-1:200, ELISA
Monoclonal
Purified
RUO
Human
GS2rab-27, GTP-binding protein Ram, HsT18676, Rab-27, RAB27A, member RAS oncogene family, RAB27MGC117246, RAMras-related protein Rab-27A
A synthesized peptide derived from Human Rab27a [UniProt P51159]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
2E8
Flow Cytometry, ELISA
Unconjugated
Rabbit
Signal Transduction
PBS, pH 7.4, 150mM NaCl and 50% glycerol
5873
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.