missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab27a Antibody (2E8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26353-50ul
This item is not returnable.
View return policy
Description
Rab27a Monoclonal antibody specifically detects Rab27a in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| Rab27a | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 2E8 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| GS2rab-27, GTP-binding protein Ram, HsT18676, Rab-27, RAB27A, member RAS oncogene family, RAB27MGC117246, RAMras-related protein Rab-27A | |
| A synthesized peptide derived from Human Rab27a [UniProt P51159] | |
| 50 μL | |
| Signal Transduction | |
| 5873 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction