missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£111.18 - £274.38
Specifications
| Antigen | RAB20 |
|---|---|
| Concentration | 0.29 mg/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
11616-1-AP is a rabbit polyclonal antibody raised against full-length of human RAB20.
Plays a role in apical endocytosis/recycling.Specifications
| RAB20 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| P35295, Q9NX57 | |
| 19332, 55647, 689377 | |
| RAB20 Fusion Protein Ag2193 | |
| Primary | |
| -20°C | |
| RAB20 |
| 0.29 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| RAB20, Ras related protein Rab 20 | |
| RAB20 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title