missing translation for 'onlineSavingsMsg'
Learn More

RAB20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16858813 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16858813 20 μL 20µL
16848813 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16858813 Supplier Proteintech Supplier No. 116161AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

11616-1-AP is a rabbit polyclonal antibody raised against full-length of human RAB20.

Plays a role in apical endocytosis/recycling.

Specifications

Antigen RAB20
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RAB20
Gene Accession No. P35295, Q9NX57
Gene Alias RAB20, Ras related protein Rab 20
Gene Symbols RAB20
Host Species Rabbit
Immunogen RAB20 Fusion Protein Ag2193
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19332, 55647, 689377
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.