missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | RAB10 |
|---|---|
| Concentration | 0.27 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16819133
|
Proteintech
11808-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16809133
|
Proteintech
11808-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.Specifications
| RAB10 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| P35281, P61026, P61027 | |
| 10890, 19325, 50993 | |
| RAB10 Fusion Protein Ag2473 | |
| Primary | |
| -20°C | |
| RAB10 |
| 0.27 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| RAB10, Ras related protein Rab 10 | |
| Rab10 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title