missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11808-1-AP-150UL
This item is not returnable.
View return policy
Description
RAB10 belongs to the RAS superfamily of small GTPases. RAB proteins localize to exocytic and endocytic compartments and regulate intracellular vesicle trafficking.Specifications
| RAB10 | |
| Polyclonal | |
| Unconjugated | |
| RAB10 | |
| RAB10, Ras related protein Rab 10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10890, 19325, 50993 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P35281, P61026, P61027 | |
| Rab10 | |
| RAB10 Fusion Protein Ag2473 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction