missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB10 Antibody (4E2), Novus Biologicals™
Mouse Monoclonal Antibody
£179.00
Specifications
| Antigen | RAB10 |
|---|---|
| Clone | 400 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
RAB10 Monoclonal specifically detects RAB10 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| RAB10 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 10890 | |
| Purified recombinant fragment of human Rab10 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 400 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| GTP-binding protein RAB10, RAB10, member RAS oncogene family, ras-related GTP-binding protein, ras-related protein Rab-10 | |
| RAB10 | |
| IgG1 | |
| Unpurified | |
| 22.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title