missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB10 Antibody (4E2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51634-0.025ml
This item is not returnable.
View return policy
Description
RAB10 Monoclonal specifically detects RAB10 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| RAB10 | |
| Monoclonal | |
| Unconjugated | |
| GTP-binding protein RAB10, RAB10, member RAS oncogene family, ras-related GTP-binding protein, ras-related protein Rab-10 | |
| Mouse | |
| 22.5 kDa | |
| 0.025 mL | |
| Signal Transduction | |
| 10890 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 400 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| RAB10 | |
| Purified recombinant fragment of human Rab10 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction