missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ R9BP Recombinant Rabbit Monoclonal Antibody (2Q8N4)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | R9BP |
|---|---|
| Clone | 2Q8N4 |
| Concentration | 1.26 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30283241
|
Invitrogen™
MA558089 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283503
![]() |
Invitrogen™
MA558090 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: MAREECKALL DGLNKTTACY HHLVLTVGGS ADSQNLRQEL QKTRQKAQEL AVSTCARLTA VLRDRGLAAD ERAEFERLWV AFSGCLDLLE ADMRRALELG AAFPLHA.
The protein encoded by this gene functions as a regulator of G protein-coupled receptor signaling in phototransduction. Studies in bovine and mouse show that this gene is expressed only in the retina, and is localized in the rod outer segment membranes. This protein is associated with a heterotetrameric complex, specifically interacting with the regulator of G-protein signaling 9, and appears to function as the membrane anchor for the other largely soluble interacting partners. Mutations in this gene are associated with prolonged electroretinal response suppression (PERRS), also known as bradyopsia.Specifications
| R9BP | |
| 1.26 mg/mL | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q6ZS82 | |
| 388531 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-107 of human R9AP/RGS9BP (NP_997274.2). | |
| Antibody |
| 2Q8N4 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| RGS9BP | |
| membrane anchor R9AP; PERRS; R9AP; regulator of G protein signaling 9 binding protein; regulator of G-protein signaling 9 binding protein; regulator of G-protein signaling 9-binding protein; regulator of G-protein signalling 9 binding protein; RGD1561747; RGS9; RGS9 anchor protein; RGS9-1 anchor; Rgs9-1bp; RGS9-anchoring protein; RGS9BP | |
| RGS9BP | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title