missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ R9BP Recombinant Rabbit Monoclonal Antibody (2Q8N4)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558089

 View more versions of this product

Product Code. 30283241

  • £147.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAREECKALL DGLNKTTACY HHLVLTVGGS ADSQNLRQEL QKTRQKAQEL AVSTCARLTA VLRDRGLAAD ERAEFERLWV AFSGCLDLLE ADMRRALELG AAFPLHA.

The protein encoded by this gene functions as a regulator of G protein-coupled receptor signaling in phototransduction. Studies in bovine and mouse show that this gene is expressed only in the retina, and is localized in the rod outer segment membranes. This protein is associated with a heterotetrameric complex, specifically interacting with the regulator of G-protein signaling 9, and appears to function as the membrane anchor for the other largely soluble interacting partners. Mutations in this gene are associated with prolonged electroretinal response suppression (PERRS), also known as bradyopsia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

R9BP
Recombinant Monoclonal
1.26 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q6ZS82
RGS9BP
Recombinant fusion protein containing a sequence corresponding to amino acids 1-107 of human R9AP/RGS9BP (NP_997274.2).
20 μL
Primary
Human
Antibody
IgG
ELISA, Immunocytochemistry
2Q8N4
Unconjugated
RGS9BP
membrane anchor R9AP; PERRS; R9AP; regulator of G protein signaling 9 binding protein; regulator of G-protein signaling 9 binding protein; regulator of G-protein signaling 9-binding protein; regulator of G-protein signalling 9 binding protein; RGD1561747; RGS9; RGS9 anchor protein; RGS9-1 anchor; Rgs9-1bp; RGS9-anchoring protein; RGS9BP
Rabbit
Protein A
RUO
388531
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.