missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-Hydroxy Bupropion, TRC

Product Code. 30390245
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390245 5 mg -
30397621 10 mg 10mg
30405849 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390245 Supplier Toronto Research Chemicals Supplier No. TRCH8306655MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 192374-15-5
Molecular Formula C13 H18 Cl N O2
Molecular Weight (g/mol) 255.74
Synonym (2R,3R)-2-(3-Chlorophenyl)-3,5,5-trimethyl-2-morpholinol, (2R-cis)-2-(3-Chlorophenyl)-3,5,5-trimethyl-2-morpholinol, (-)-(2R,3R)-2-(3-Chlorophenyl)-3,5,5-trimethyl-2-morpholinol
IUPAC Name (2R,3R)-2-(3-chlorophenyl)-3,5,5-trimethylmorpholin-2-ol
SMILES C[C@H]1NC(C)(C)CO[C@]1(O)c2cccc(Cl)c2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H18ClNO2/c1-9-13(16,17-8-12(2,3)15-9)10-5-4-6-11(14)7-10/h4-7,9,15-16H,8H2,1-3H3/t9-,13+/m1/s1
Formula Weight 255.1026
Chemical Name or Material (R,R)-hydroxy Bupropion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.