CAS RN 192374-15-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R,R)-Hydroxy Bupropion, TRC
CAS: 192374-15-5 Molecular Formula: C13 H18 Cl N O2 Molecular Weight (g/mol): 255.74 Synonym: (2R,3R)-2-(3-Chlorophenyl)-3,5,5-trimethyl-2-morpholinol,(2R-cis)-2-(3-Chlorophenyl)-3,5,5-trimethyl-2-morpholinol,(-)-(2R,3R)-2-(3-Chlorophenyl)-3,5,5-trimethyl-2-morpholinol IUPAC Name: (2R,3R)-2-(3-chlorophenyl)-3,5,5-trimethylmorpholin-2-ol SMILES: C[C@H]1NC(C)(C)CO[C@]1(O)c2cccc(Cl)c2