missing translation for 'onlineSavingsMsg'
Learn More

(R)-De(carboxymethyl) Cetirizine Ethanol Dihydrochloride, TRC

Product Code. 30009047
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009047 1 g 1g
30009018 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009047 Supplier Toronto Research Chemicals Supplier No. TRCD2155101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 728948-88-7
Molecular Formula C19 H23 Cl N2 O . 2 Cl H
Molecular Weight (g/mol) 403.77
Synonym 1-Piperazineethanol, 4-[(R)-(4-chlorophenyl)phenylmethyl]-, hydrochloride (1:2, 1-Piperazineethanol, 4-[(R)-(4-chlorophenyl)phenylmethyl]-, dihydrochloride (9CI), 2-[4-[(R)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethanol dihydrochloride
IUPAC Name 2-[4-[(R)-(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethanol;dihydrochloride
SMILES Cl.Cl.OCCN1CCN(CC1)[C@H](c2ccccc2)c3ccc(Cl)cc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H23ClN2O.2ClH/c20-18-8-6-17(7-9-18)19(16-4-2-1-3-5-16)22-12-10-21(11-13-22)14-15-23;;/h1-9,19,23H,10-15H2;2*1H/t19-;;/m1../s1
Formula Weight 402.1032
Chemical Name or Material (R)-De(carboxymethyl) Cetirizine Ethanol Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.