CAS RN 728948-88-7
CAS RN 728948-88-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-De(carboxymethyl) Cetirizine Ethanol Dihydrochloride, TRC
CAS: 728948-88-7 Molecular Formula: C19 H23 Cl N2 O . 2 Cl H Molecular Weight (g/mol): 403.77 Synonym: 1-Piperazineethanol, 4-[(R)-(4-chlorophenyl)phenylmethyl]-, hydrochloride (1:2,1-Piperazineethanol, 4-[(R)-(4-chlorophenyl)phenylmethyl]-, dihydrochloride (9CI),2-[4-[(R)-(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethanol dihydrochloride IUPAC Name: 2-[4-[(R)-(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethanol;dihydrochloride SMILES: Cl.Cl.OCCN1CCN(CC1)[C@H](c2ccccc2)c3ccc(Cl)cc3