missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ TACS-XL In Situ Apoptosis Detection Kits
Description
Includes
Proteinase K Solution; Cytonin; TdT enzyme, B-dNTP mix; Strep-HRP; anti-BrdU; Strep-Diluent
Based on incorporation of bromodeoxyuridine (BrdU) at the 3 ft. OH ends of the DNA fragments that form during apoptosis
This detection system uses a biotin conjugated anti-BrdU antibody and streptavidin-horseradish peroxidase for visualiza
Specifications
Specifications
| Quantity | 30 Tests |
| Product Type | TACS-XL In Situ Apoptosis Detection Kit |
| Content And Storage | The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| Detection Method | DAB |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?