missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ TACS TdT In Situ Apoptosis Detection Kit, Blue Label

Product Code. 18411560
Change view
Click to view available options
Detection Method:
Blue Label
DAB
Unit Size:
30 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detection Method unitSize
18411560 Blue Label 30 tests
18432620 DAB 30 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18411560 Supplier R&D Systems™ Supplier No. 481130K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Proteinase K Solution; Cytonin™; TdT enzyme, dNTP mix and buffers; Strep-HRP; TACS-Blue Label; Blue Streptavidin-Diluent; Cations; TACS-Nuclease™ and buffer; Nuclear Fast Red

A TUNEL assay designed for fixed cells, embedded tissue, and frozen tissue using light and electron microscopy

Contains a highly purified form of the TdT enzyme for the enzymatic incorporation of biotinylated nucleotides
Biotin labeling is achieved using streptavidin-horseradish peroxidase and the colorimetric substrate TACS Blue Label

TRUSTED_SUSTAINABILITY

Specifications

Quantity 30 Tests
Kit Contents Proteinase K, Cytonin, TdT enzyme, dNTPs, StrepHRP, TACS-Blue Label, Blue Streptavidn-Diluent, Cations, TACS-Nuclease, Nuclear Fast Red
Product Type TACS TdT In Situ Apoptosis Detection Kit
Content And Storage The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Detection Method Blue Label
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.