Learn More
R&D Systems™ TACS Annexin V-FITC Apoptosis Detection Kit
Description
Includes
TACS Annexin V-FITC, 250uL; 10X Binding Buffer, 20mL; Propidium Iodide, 2.5mL
Early in the apoptosis process, phosphatidylserine (PS) becomes exposed on the cell surface by flipping from the inner to outer leaflet of the cytoplasmic membrane. This event is thought to be important for macrophage recognition of cells undergoing apopt
Supplied with an optimized binding buffer and propidium iodide, included in the incubation mix to identify cells that have lost membrane integrity
As cells disintegrate, greater access to the inner cell membrane allows for additional Annexin V binding: T
Specifications
Specifications
| Quantity | 250 Tests |
| Kit Contents | TACS Annexin V-FITC, 250μL; 10X Binding Buffer, 20mL; Propidium Iodide, 2.5mL |
| Product Type | TACS Annexin V-FITC Apoptosis Detection Kit |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| For Use With (Application) | Facilitates rapid fluorometric quantitation of apoptotic cells |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.