missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Wnt-3a (High Purity) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Specifications
Gene ID (Entrez) | 22416 |
---|---|
Molecular Weight (g/mol) | 37.4 kDa |
Name | Wnt-3a (High Purity) |
Quantity | 10μg |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-3a protein Ser19-Lys352 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18375547
|
R&D Systems™
1324-WNP-010 |
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS with BSA as a carrier protein. |
N/A
|
N/A | |||||
18310548
|
R&D Systems™
1324-WNP-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS, NaCl, EDTA and CHAPS. |
N/A
|
N/A | |||||
Description
The Recombinant Mouse Wnt-3a (High Purity) Protein is derived from CHO. The Recombinant Mouse Wnt-3a (High Purity) Protein has been validated for the following applications: Bioactivity.Specifications
22416 | |
Wnt-3a (High Purity) | |
Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-3a protein Ser19-Lys352 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
37.4 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |