missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ MagCellect Human Memory CD4+ T Cell Isolation Kit

Product Code. 18472190
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18472190

Brand: R&D Systems™ MAGH116

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed to isolate CD4+ Memory T cells via a negative selection principle

CD4 is an approximately 55kDa type I transmembrane glycoprotein that is expressed predominantly on thymocytes and a subset of mature T cells. In humans, CD4 is also expressed to a lesser extent on monocytes and macrophage related cells.

Typical purity of recovered CD4+ Memory T cells ranges between 75% and 85%
Kit protocols based on the use of Ferrofluids or magnetic nanoparticles that have no magnetic memory (superparamagnetic)
These Ferrofluids have a diameter of 150nm and as a result behave like colloidal particles, offering a higher ligand capacity per mass compared to other, larger magnetic particles

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Quantity 1 Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.