missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-3b/GDF-10 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£526.98
Specifications
Gene ID (Entrez) | 2662 |
---|---|
Molecular Weight (g/mol) | 12.5 kDa (monomer) |
Name | BMP-3b/GDF-10 |
Quantity | 25μg |
Source | E. coli-derived human BMP-3b/GDF-10 protein Gln369-Arg478,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18339667
|
R&D Systems™
1543-BP-025 |
£526.98
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18116590
|
R&D Systems™
1543-BP-025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
£526.98
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human BMP-3b/GDF-10 Protein is derived from E. coli. The Recombinant Human BMP-3b/GDF-10 Protein has been validated for the following applications: Bioactivity.Specifications
2662 | |
BMP-3b/GDF-10 | |
E. coli-derived human BMP-3b/GDF-10 protein Gln369-Arg478,with an N-terminal Met | |
BIP, BMP-3b, BMP3B, bone morphogenetic protein 3B, Bone-inducing protein, GDF10, GDF-10, growth differentiation factor 10, Growth/differentiation factor 10 | |
Recombinant |
12.5 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |