missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-3b/GDF-10 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1543-BP-025/CF
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human BMP-3b/GDF-10 Protein is derived from E. coli. The Recombinant Human BMP-3b/GDF-10 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | |
12.5 kDa (monomer) | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
BIP, BMP-3b, BMP3B, bone morphogenetic protein 3B, Bone-inducing protein, GDF10, GDF-10, growth differentiation factor 10, Growth/differentiation factor 10 | |
Recombinant |
2662 | |
BMP-3b/GDF-10 | |
E. coli-derived human BMP-3b/GDF-10 protein Gln369-Arg478,with an N-terminal Met | |
Disulfide-linked homodimer | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |