missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ HT Superoxide Dismutase Assay Kit

Product Code. 18402250
Change view
Click to view available options
Quantity:
480 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402250 480 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402250 Supplier R&D Systems™ Supplier No. 7501500K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

SOD, SOD Reaction Buffer, Xanthine Solution, Xanthine Oxidase Solution, Triton™ X-100, WST-1 Reagent, Five 96-well plates for 480 tests

A 96-well microplate-based assay designed for high-throughput analysis of Superoxide Dismutase (SOD) activity in mammalian tissue and cell lysates

SODs are metalloenzymes that catalyze the dismutation of the superoxide radical into hydrogen peroxide and molecular oxygen, providing an important defense against oxidative damage.

Free of interference by other catalytic activities and well-suited to assaying SOD in mammalian cell lysates
Not greatly disturbed by trace metals
Performed in as little as 6 minutes
The superoxide anion then coverts the tetrazolium salt WST-1 to the c

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Quantity 480 Tests
compliance-icons
Product Identifier
  • HT Superoxide Dismutase Assay Kit
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Mixture of 5-Chloro-2-Methyl-4- Isothiazolin-3-One
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.