Learn More
R&D Systems™ Cultrecoat High BME Cell Invasion Assay, 96-well
A ready-to-use assay for cell invasion into a medium density extracellular matrix.
POA
Specifications
Quantity 1 Kit
View More Specs
Includes: 96 Well BME Cell Invasion Chamber (Low, Medium or High); 25X Cell Wash Buffer, 2 x 1.5mL; 10X Cell Dissociation Solution, 2 x 1.5mL; Calcein-AM, 50ug
Description
Description
Cell invasion is fundamental to angiogenesis, embryonic development, immune responses and tumor cell metastasis.
Contains one 96-well plate that is coated with BME, for cell models that exhibit a nominal invasive capacity: The low amount of BME coating provides the highest sensitivity for minimally invasive cell models
The medium amount of BME coating provides the
Specifications
Specifications
1 Kit