missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(Boc-amino)-4-(4-bromophenyl)butyric acid, 95%, Thermo Scientific™

Product Code. 15499615
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15499615

Brand: Thermo Scientific Alfa Aesar H52136.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-3-(Boc-amino)-4-(4-bromophenyl)butyric acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 331763-75-8
Molecular Formula C15H20BrNO4
Molecular Weight (g/mol) 358.232
MDL Number MFCD01860917
InChI Key GYXZKHKUVVKDHI-GFCCVEGCSA-N
Synonym r-4-4-bromophenyl-3-tert-butoxycarbonyl amino butanoic acid, 3r-4-4-bromophenyl-3-tert-butoxycarbonyl amino butanoic acid, boc-d-?-homophe 4-br-oh, boc-4-bromo-d-b-homophenylalanine, 3r-4-4-bromophenyl-3-tert-butoxycarbonylamino butanoic acid, r-4-4-bromophenyl-3-tert-butoxycarbonylamino butanoic acid, benzenebutanoic acid, 4-bromo-beta-1,1-dimethylethoxy carbonyl amino-, betar
PubChem CID 7009737
IUPAC Name (3R)-4-(4-bromophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)Br)CC(=O)O

Specifications

Melting Point 144°C to 146°C
Quantity 1 g
Formula Weight 358.23
Percent Purity 95%
Chemical Name or Material (R)-3-(Boc-amino)-4-(4-bromophenyl)butyric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.