accessibility menu, dialog, popup

UserName

CAS RN 331763-75-8

CH3CH3H3COOHNBrOOH

CAS RN 331763-75-8

IUPAC Name: 4-(4-bromophenyl)-3-{[(tert-butoxy)carbonyl]amino}butanoic acid
Synonyms: 4-(4-bromophenyl)-3-[(tert-butoxycarbonyl)amino]butanoic acid
Molecular Weight (g/mol): 358.23
Molecular Formula: C15H20BrNO4
InChi Key: GYXZKHKUVVKDHI-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CC(O)=O)CC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)