missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Isopropylsuccinic acid-1-methyl ester, 95%, (98% ee), Thermo Scientific Chemicals

Product Code. 10350652 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10350652 500 mg Glass bottle 500mg
10792095 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10350652 Supplier Thermo Scientific Acros Supplier No. 363065000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220498-08-8
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.19
MDL Number MFCD01091030
InChI Key VHOGSWBAPDPWTG-ZCFIWIBFSA-N
Synonym r-3-methoxycarbonyl-4-methylpentanoic acid, r-2-isopropyl-succinic acid-1-methyl ester, r-2-isopropylsuccinic acid-1-methyl ester, 3r-3-methoxycarbonyl-4-methylpentanoic acid, 2r-2-isopropylsuccinic acid 1-methyl ester, r---2-isopropylsuccinic acid 1-methyl ester, butanedioic acid,2-1-methylethyl-, 1-methyl ester, 2r
PubChem CID 5702634
IUPAC Name (3R)-3-methoxycarbonyl-4-methylpentanoic acid
SMILES CC(C)C(CC(=O)O)C(=O)OC

Specifications

Color Yellow
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500 mg
Formula Weight 174.19
Percent Purity 95%,(98% ee)
Physical Form Viscous Liquid
Chemical Name or Material (R)-2-Isopropylsuccinic acid-1-methyl ester, 98% ee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.