accessibility menu, dialog, popup

UserName

CAS RN 220498-08-8

H3COO(R)OOCH3H3C

CAS RN 220498-08-8

IUPAC Name: (3R)-3-(methoxycarbonyl)-4-methylpentanoate
Synonyms: (3R)-3-(methoxycarbonyl)-4-methylpentanoate
Molecular Weight (g/mol): 173.19
Molecular Formula: C8H13O4
InChi Key: VHOGSWBAPDPWTG-ZCFIWIBFSA-M
SMILES: COC(=O)[C@H](CC([O-])=O)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results