missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,2-Propanediol, 98%

Product Code. 11425037
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11425037 25 g 25g
11405037 1 g 1g
11415037 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425037 Supplier Thermo Scientific Alfa Aesar Supplier No. B21937.14
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4254-14-2 | C3H8O2 | 76.095 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4254-14-2
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00066248
InChI Key DNIAPMSPPWPWGF-GSVOUGTGSA-N
Synonym r---1,2-propanediol, r-propane-1,2-diol, 2r-propane-1,2-diol, r-1,2-propanediol, r-propylene glycol, 2r-1,2-propanediol, r---1,2-dihydroxypropane, unii-602hn5l69h
PubChem CID 259994
ChEBI CHEBI:28972
IUPAC Name (2R)-propane-1,2-diol
SMILES CC(CO)O

Specifications

Density 1.04
Boiling Point 186°C to 188°C
Flash Point 107°C (224°F)
Refractive Index 1.432
Quantity 25 g
Beilstein 1718872
Sensitivity Hygroscopic
Merck Index 14,7855
Optical Rotation −16.5° (Neat)
Formula Weight 76.1
Percent Purity 98%
Chemical Name or Material (R)-(-)-1,2-Propanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.