accessibility menu, dialog, popup

UserName

CAS RN 4254-14-2

IUPAC Name: (2R)-propane-1,2-diol
Synonyms: R-1,2-propanediol S-1,2-propanediol 2,3-propandiol
Molecular Weight (g/mol): 76.1
Molecular Formula: C3H8O2
InChi Key: DNIAPMSPPWPWGF-GSVOUGTGSA-N
SMILES: C[C@@H](O)CO
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results