missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinazoline, 98%, Thermo Scientific Chemicals
£29.70 - £108.00
Chemical Identifiers
CAS | 253-82-7 |
---|---|
Molecular Formula | C8H6N2 |
Molecular Weight (g/mol) | 130.15 |
MDL Number | MFCD00006712 |
InChI Key | JWVCLYRUEFBMGU-UHFFFAOYSA-N |
Synonym | phenmiazine, 1,3-benzodiazine, 1,3-diazanaphthalene, 5,6-benzopyrimidine, benzo a pyrimidine, chinazolin, unii-ub9qur18nl, ub9qur18nl, benzopyrimidine |
PubChem CID | 9210 |
ChEBI | CHEBI:36621 |
IUPAC Name | quinazoline |
SMILES | C1=CC=C2N=CN=CC2=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
253-82-7 | |
130.15 | |
JWVCLYRUEFBMGU-UHFFFAOYSA-N | |
9210 | |
quinazoline |
C8H6N2 | |
MFCD00006712 | |
phenmiazine, 1,3-benzodiazine, 1,3-diazanaphthalene, 5,6-benzopyrimidine, benzo a pyrimidine, chinazolin, unii-ub9qur18nl, ub9qur18nl, benzopyrimidine | |
CHEBI:36621 | |
C1=CC=C2N=CN=CC2=C1 |
Specifications
253-82-7 | |
Brown to Yellow | |
106°C | |
97.5% min. (GC) | |
MFCD00006712 | |
15, 8169 | |
JWVCLYRUEFBMGU-UHFFFAOYSA-N | |
quinazoline | |
9210 | |
130.15 | |
Crystalline Powder, Crystals or Chunks |
45°C to 48°C | |
243°C | |
Authentic | |
C8H6N2 | |
23, 175 | |
phenmiazine, 1,3-benzodiazine, 1,3-diazanaphthalene, 5,6-benzopyrimidine, benzo a pyrimidine, chinazolin, unii-ub9qur18nl, ub9qur18nl, benzopyrimidine | |
C1=CC=C2N=CN=CC2=C1 | |
130.15 | |
CHEBI:36621 | |
98% | |
Quinazoline, 98% |
Safety and Handling
missing translation for 'einecsNumber' : 205-965-3
RUO – Research Use Only