missing translation for 'onlineSavingsMsg'
Learn More

Quinazoline, 98%

Product Code. 10436990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10436990 1 g Glass bottle 1g
10446990 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10436990 Supplier Thermo Scientific Acros Supplier No. 153980010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 253-82-7 | C8H6N2 | 130.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253-82-7
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00006712
InChI Key JWVCLYRUEFBMGU-UHFFFAOYSA-N
Synonym phenmiazine, 1,3-benzodiazine, 1,3-diazanaphthalene, 5,6-benzopyrimidine, benzo a pyrimidine, chinazolin, unii-ub9qur18nl, ub9qur18nl, benzopyrimidine
PubChem CID 9210
ChEBI CHEBI:36621
IUPAC Name quinazoline
SMILES C1=CC=C2N=CN=CC2=C1

Specifications

Melting Point 45°C to 48°C
Color Brown to Yellow
Boiling Point 243°C
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 23, 175
Merck Index 15, 8169
Formula Weight 130.15
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Quinazoline, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.