missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Vigreux Fractionating Column

Vigreux type with combination of horizontal and angled indentations.

Brand:  Quickfit FC3/23

Code : NEW

Additional Details : Weight : 0.26000kg

 View more versions of this product

Product Code. 15537820

  • £1050.00 / Pack of 5
Estimated Shipment: 05-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Popular, easy to use column
  • Vigreux type with combination of horizontal and angled indentations
  • Horizontal indentations ensure intimate vapour and liquid contact
  • Angled indentations redistribute liquid from walls to center of column
Specifications

Specifications

600 mm
Vigreux Fractionating Column
ISO 3585
600mm
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers