Learn More
Quickfit™ Borosilicate Glass Vigreux Fractionating Column
Product Code. p-8002936
Change view
Click to view available options
Height (Metric):
360 mm
600 mm
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
15527820 360 mm Pack of 5
15537820 600 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15527820 Supplier Quickfit Supplier No. FC3/14
Only null left
Add to basket
Add to basket
Vigreux type with combination of horizontal and angled indentations.
  • Popular, easy to use column
  • Vigreux type with combination of horizontal and angled indentations
  • Horizontal indentations ensure intimate vapour and liquid contact
  • Angled indentations redistribute liquid from walls to center of column
TRUSTED_SUSTAINABILITY
Specifications
Includes 2 NS 29/32 standard ground joint
Height (Metric) 360 mm
Length (Metric) 360mm
Type Vigreux Column
Material Glass
Quantity 1Each
Standard Taper Joint NS 29/32