Learn More
DWK Life Sciences™ Quick Release Clamp, from Stainless steel, with retaining clip
Tienda DWK Life Sciences Productos
missing translation for 'orderingAttributeHoverText'
Size:
DN 100
DN 120
DN 150
DN 200
DN 60
missing translation for 'unitSize'
Each
Descripción
The quick release clamp closes flat flange vessels and secures the lid. Slipping or unchecked opening is not possible. The closure is made of high-quality Stainless steel and has three holding segments.
- With retaining clip
- Quick closing with quick-release fastener
Especificaciones
Especificaciones
| Quantity | 1 Ea. |
| Material | Stainless Steel |
| For Use With (Equipment) | Quick Release Clamp for Closure |
| For Use With (Application) | Quick release clamp for closure; Accessories for flat flange vessels |
| Size | DN 100 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido