Learn More
DWK Life Sciences™ Quick Release Clamp, from Stainless steel, with retaining clip
Shop All DWK Life Sciences ProductsDescription
The quick release clamp closes flat flange vessels and secures the lid. Slipping or unchecked opening is not possible. The closure is made of high-quality Stainless steel and has three holding segments.
- With retaining clip
- Quick closing with quick-release fastener
Specifications
Specifications
| Quantity | 1 Ea. |
| Material | Stainless Steel |
| For Use With (Equipment) | Quick Release Clamp for Closure |
| For Use With (Application) | Quick release clamp for closure; Accessories for flat flange vessels |
| Size | DN 150 |