Learn More
DWK Life Sciences Quick Release Clamp, from Stainless steel, with retaining clip
The quick release clamp is used to close flat flange vessels.
Brand: DWK Life Sciences 290715108
67.52 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Code : A0
Additional Details : Weight : 0.24400kg
Description
The quick release clamp closes flat flange vessels and secures the lid. Slipping or unchecked opening is not possible. The closure is made of high-quality Stainless steel and has three holding segments.
- With retaining clip
- Quick closing with quick-release fastener
Specifications
1 Ea. | |
Quick Release Clamp for Closure | |
DN 120 |
Stainless Steel | |
Quick release clamp for closure; Accessories for flat flange vessels |