Learn More
DWK Life Sciences™ Quick Release Clamp, from Stainless steel, with retaining clip
Shop All DWK Life Sciences Products
Click to view available options
Size:
DN 100
DN 120
DN 150
DN 200
DN 60
Unit Size:
Each
Description
The quick release clamp closes flat flange vessels and secures the lid. Slipping or unchecked opening is not possible. The closure is made of high-quality Stainless steel and has three holding segments.
- With retaining clip
- Quick closing with quick-release fastener
Specifications
Specifications
| Quantity | 1 Ea. |
| Material | Stainless Steel |
| For Use With (Equipment) | Quick Release Clamp for Closure |
| For Use With (Application) | Quick release clamp for closure; Accessories for flat flange vessels |
| Size | DN 60 |