missing translation for 'onlineSavingsMsg'
Learn More

Quercetin hydrate, 95%

Product Code. 10614862 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10614862 100 g Glass bottle 100g
10484375 10 g Glass bottle 10g
10604862 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10614862 Supplier Thermo Scientific Acros Supplier No. 174071000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 849061-97-8 | C15H10O7 | 302.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 849061-97-8
Molecular Formula C15H10O7
Molecular Weight (g/mol) 302.24
MDL Number MFCD03847906
InChI Key REFJWTPEDVJJIY-UHFFFAOYSA-N
Synonym quercetin hydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, quercetinhydrate, quercetine sophoretin /, regid_for_cid_747934, regid_for_cid_16212154, 3,3;,4;,5,7-pentahydroxyflavon, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate
PubChem CID 16212154
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
SMILES OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1

Specifications

Melting Point >300°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Beilstein 18, 242
Merck Index 14, 8034
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in ethanol,acetone,pyridine,,glacial acetic acid
Formula Weight 302.24
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Quercetin hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Quercetin hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.