missing translation for 'onlineSavingsMsg'
Learn More

Quercetin dihydrate, 99.85%, For HPLC analysis, MP Biomedicals™

Product Code. 11424281 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424281 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424281 Supplier MP Biomedicals Supplier No. 0215200310
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6151-25-3
Molecular Formula C15H14O9
Molecular Weight (g/mol) 338.27
MDL Number MFCD00149487
InChI Key GMGIWEZSKCNYSW-UHFFFAOYSA-N
Synonym quercetin dihydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one dihydrate, quercetine dihydrate, quercetin, dihydrate, ccris 3304, 3,3',4',5,7-pentahydroxyflavone dihydrate, quercetin dihydrate sophoretin, flavone, 3,3',4',5,7-pentahydroxy-, dihydrate, dsstox_cid_1219, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-1-benzopyran-4-one dihydrate
PubChem CID 5284452
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate
SMILES O.O.OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1

Specifications

Color Green-Yellow
Quantity 10 g
Formula Weight 338.26
Percent Purity 0.9985
Physical Form Powder
Chemical Name or Material Quercetin dihydrate, Molecular Biology
compliance-icons
Product Identifier
  • Quercetin Dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P330-Rinse mouth.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 3,3',4',5,7-Pentahydroxyflavone dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.