missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Quantofix™ Zinc Test Strip Kits

Product Code. 10411591 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color Indicator:
Orange to Red
Unit Size:
81g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Indicator unitSize
10411591 Orange to Red 81g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10411591 Supplier Macherey-Nagel™ Supplier No. 91310

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allows for the quick and easy determination of zinc in solutions. Macherey Nagel™ Quantofix™ Zinc Test Strip Kits come pre-calibrated and contain all necessary equipment and reagents.

  • Rapid: most tests can be carried out in 10 to 120s
  • Excellent for checking plating baths in the metal plating industry
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C

Specifications

Color Indicator Orange to Red
Format Strip
For Use With (Application) Determination of Zinc
Graduations 0, 2, 5, 10, 25, 50, 100mg/L Zn
Quantity 100 Pack
Storage Requirements 30°C
Test Time 2 min.
Type Semi-qualitative Test Strip
compliance-icons
Product Identifier
  • QUANTOFIX Zinc
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.