missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Quantofix™ Tin Semi-quantitative Test Strips
Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
White to Dark Blue
Unit Size:
Pack of 100
Description
- Rapid: most tests can be carried out in 10s to 120s
- Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
- Precise: color charts adjusted and checked using certified standard solutions
- Shelf life: 2. 5 years, store below 30°C
Specifications
Specifications
| Color Indicator | White to Dark Blue |
| Format | Strip |
| For Use With (Application) | Determination of Tin |
| Graduations | 0, 10, 25, 50, 100, 250, 500mg/L Sn2+ |
| Quantity | 100 Pack |
| Storage Requirements | 30°C |
| Test Time | 5 sec. |
| Type | Semi-qualitative Test Strip |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction