missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ QUANTOFIX ™ Semi-quantitative Nitrate Test Strips

Product Code. 15747958 Shop All Macherey-Nagel Products
Click to view available options
Quantity:
100 Strips
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15747958

Brand: Macherey-Nagel™ 91351

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Semi-quantitative test strips for the determination of Nitrate and Nitrite. Ideal for rapid and easy analysis of Nitrate and Nitrite directly on-site. Can be used with test strip reader QUANTOFIX Relax.

  • Ideal for detemination of nitrate and nitrite
  • Use with QUANTOFIX Relax reflectometer
  • 2.5 year shelf life

Specifications

Color Yellow to Red/Violet
Format 10 Test Strips/Tube
For Use With (Application) Nitrate and Nitrite Testing. Nitrate - Modified Griess Reaction, Nitrite - Griess Reaction
Includes Instruction Leaflet
No. of Tests 100 Strips
Quantity 100 Strips
Range Nitrate: 3 to 100 mg/L NO3-, Nitrite: 0.5 to 50 mg/L NO2-
Storage Requirements 4°C to 30°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.