missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Quantofix™ Nickel Test Strip Kits

Product Code. 10100491 Shop All Macherey-Nagel Products
Click to view available options
Color Indicator:
White to Bright Red
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10100491

Brand: Macherey-Nagel™ 91305

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Allows for the quick and easy determination of nickel in solutions. Macherey Nagel™ Quantofix™ Nickel Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.

  • Ready to use
  • Rapid: provides reliable results within 30 seconds
  • Excellent for use in metal plating processes and electroplating solutions
  • Precise: colour charts adjusted and checked using certified standard solutions
  • Shelf life: 2.5 years, store below 30°C

Specifications

Color Indicator White to Bright Red
Format Strip
For Use With (Application) Determination of Nickel
Graduations 0, 10, 25, 50, 100, 250, 500, 1000mg/L Ni2+
Quantity 100 Pack
Storage Requirements 30°C
Test Time 30 sec.
Type Semi-qualitative Test Strip
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.