missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
- Rapid: most tests can be carried out in 10s to 120s
- Convenient: ready to use kits, pre-calibrated, contain all necessary equipment and reagents
- Precise: color charts adjusted and checked using certified standard solutions
- Shelf life: 2. 5 years, store below 30°C
Specifications
Specifications
| Color Indicator | White to Violet |
| Format | Strip |
| For Use With (Application) | Determination of Cyanide |
| Graduations | 0, 1, 3, 10, 30mg/L CN- |
| Quantity | 100 Pack |
| Storage Requirements | 30°C |
| Test Time | 3 min. |
| Type | Semi-qualitative Test Strip |
Safety and Handling
Product Identifier
- QUANTOFIX Cyanide
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : pyridine, chloramineT
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?