missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Allows for the quick and easy determination of copper in solutions. Macherey Nagel™ Quantofix™ Copper Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
- Ready to use
- Rapid: provides reliable results within 20 seconds
- Excellent for the electroplating process and water sampling
- Precise: colour charts adjusted and checked using certified standard solutions
- Self life: 2.5 years, store below 30°C
Specifications
Specifications
| Color Indicator | White to Red/Violet |
| Format | Strip |
| For Use With (Application) | Determination of Copper |
| Graduations | 0, 10, 30, 100, 300mg/L Cu1+/2+ |
| Quantity | 100 Pack |
| Storage Requirements | 30°C |
| Test Time | 20 sec. |
| Type | Semi-qualitative Test Strip |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?